Skip to Content
Merck
All Photos(1)

Key Documents

24233

Sigma-Aldrich

Dichloromethane

puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)

Synonym(s):

methylene chloride, Methylene chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$79.00
2.5 L
$107.00
250 KG
Inquire

$79.00


Estimated to ship on21 March 2025


Request a Bulk Order

Select a Size

Change View
1 L
$79.00
2.5 L
$107.00
250 KG
Inquire

About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.04
Assay:
≥99% (GC)
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

$79.00


Estimated to ship on21 March 2025


Request a Bulk Order

vapor density

2.9 (vs air)

Quality Level

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

grade

puriss.

Assay

≥99% (GC)

form

liquid

autoignition temp.

1223 °F

quality

meets analytical specification of Ph. Eur., NF

contains

10-60 ppm amylene as stabilizer

expl. lim.

22 %

impurities

residual solvents, complies
≤0.0001% free chlorine (Cl)
≤0.001% carbon tetrachloride (GC)
≤0.001% free acid (as HCl)
≤0.002% non-volatile matter
≤0.005% trichloromethane (GC)
≤0.02% methanol (GC)
≤0.02% water (Karl Fischer)
≤1 ppm heavy metals (as Pb)
≤2.0% ethanol (GC)

evapn. residue

≤20 ppm

refractive index

n20/D 1.423-1.425
n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.318-1.322 g/mL at 25 °C
1.325 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤3 mg/kg

suitability

complies for appearance of solution
complies for identity (IR)
complies for reaction against H2SO4

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane has been tested as a solvent medium for the dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml.[1] Role of the quantity of TiO2 loading on activated carbon support on its photodecomposition has been investigated.[2]

Application

Dichloromethane may be used as solvent for the preparation of stock solutions of the carotenoids.[3] It may be employed as solvent in the synthesis of polylactide-based (PLA) thin films.[4]

Other Notes

The article number 24233-4X2.5L-M will be discontinued. Please order the single bottle 24233-2.5L-M which is physically identical with the same exact specifications.
The article number 24233-4X2.5L-R will be discontinued. Please order the single bottle 24233-2.5L-R which is physically identical with the same exact specifications.
The article number 24233-6X1L-M will be discontinued. Please order the single bottle 24233-1L-M which is physically identical with the same exact specifications.
The article number 24233-6X1L-R will be discontinued. Please order the single bottle 24233-1L-R which is physically identical with the same exact specifications.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Re et al.
Free radical biology & medicine, 26(9-10), 1231-1237 (1999-06-25)
A method for the screening of antioxidant activity is reported as a decolorization assay applicable to both lipophilic and hydrophilic antioxidants, including flavonoids, hydroxycinnamates, carotenoids, and plasma antioxidants. The pre-formed radical monocation of 2,2'-azinobis-(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS*+) is generated by oxidation
V Melissinaki et al.
Biofabrication, 3(4), 045005-045005 (2011-09-21)
This study reports on the production of high-resolution 3D structures of polylactide-based materials via multi-photon polymerization and explores their use as neural tissue engineering scaffolds. To achieve this, a liquid polylactide resin was synthesized in house and rendered photocurable via
Dipyridine-chromium (VI) oxide oxidation of alcohols in dichloromethane.
Collins JC, et al.
Tetrahedron Letters, 9(30), 3363-3366 (1968)
Effect of activated carbon content in TiO2-loaded activated carbon on photodegradation behaviors of dichloromethane.
Torimoto T, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 103(1), 153-157 (1997)
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service