Merck
  • DJ-1 mediates the resistance of cancer cells to dihydroartemisinin through reactive oxygen species removal.
DJ-1 mediates the resistance of cancer cells to dihydroartemisinin through reactive oxygen species removal.
Free radical biology & medicine (2014-04-01)
Hong Zhu, Si-Da Liao, Jia-Jie Shi, Lin-Lin Chang, Yun-Guang Tong, Ji Cao, Ying-Ying Fu, Xiu-Ping Chen, Mei-Dan Ying, Bo Yang, Qiao-Jun He, Jin-Jian Lu
ABSTRACT
Dihydroartemisinin (DHA), one of the main metabolites of artemisinin and its derivatives, presents anti-cancer potential in vitro and in vivo. To explore the mechanisms of resistance toward DHA, a DHA-resistant cell line, HeLa/DHA, was established with a resistance factor of 7.26 in vitro. Upon DHA treatment, apoptotic cells were significantly elicited in parental HeLa cells but minimally induced in HeLa/DHA cells. HeLa/DHA cells also displayed much less sensitivity to DHA-induced tumor suppression in cancer xenograft models than HeLa cells. Intriguingly, DHA-resistant cells did not display a multidrug-resistant phenotype. Based on a proteomic study employing LC-ESI-MS/MS together with pathway analysis, DJ-1 (PARK7) was found to be highly expressed in HeLa/DHA cells. Western blot and immunofluorescence assays confirmed the higher expression of DJ-1 in HeLa/DHA cells than in parental cells in both cell line and xenograft models. DJ-1 is translocated to the mitochondria of HeLa/DHA cells and oxidized, providing DJ-1 with stronger cytoprotection activity. Further study revealed that DJ-1 knockdown in HeLa/DHA cells abolished the observed resistance, whereas overexpression of DJ-1 endowed the parental HeLa cells with resistance toward DHA. Reactive oxygen species (ROS) were also significantly induced by either DHA or hydrogen peroxide in HeLa cells but not in resistant HeLa/DHA cells. When the cells were pretreated with N-acetyl-l-cysteine, the effect of DJ-1 knockdown on sensitizing HeLa/DHA cells to DHA was significantly attenuated. In summary, our study suggests that overexpression and mitochondrial translocation of DJ-1 provides HeLa/DHA cells with resistance to DHA-induced ROS and apoptosis.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Park7
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Supelco
5-Fluorouracil, analytical standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human PARK7
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Propidium iodide solution
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Etoposide, European Pharmacopoeia (EP) Reference Standard
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard