Skip to Content
Merck
All Photos(1)

Key Documents

F0250000

Fluorouracil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Fluorouracil, 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing

Select a Size

35 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
35 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
Beilstein:
127172
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

fluorouracil

manufacturer/tradename

EDQM

mp

282-286 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC1=CNC(=O)NC1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Gene Information

human ... TYMS(7298)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluorouracil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Bp van Kuilenburg et al.
Pharmacogenomics, 14(7), 799-811 (2013-05-09)
5-fluorouracil (5-FU) remains the cornerstone of all currently applied regimens for the treatment of patients with cancers of the gastrointestinal tract, breast, and head and neck. Unfortunately, a large variation in the clearance of 5-FU has been observed between patients
Tania Diaz et al.
Journal of surgical oncology, 109(7), 676-683 (2014-02-11)
Surgery is the standard treatment for colorectal cancer (CRC), and adjuvant chemotherapy has been shown to be effective in stage III but less so in stage II. We have analyzed the expression of the miR-200 family in tissue samples from
Guido Deboever et al.
Clinical colorectal cancer, 12(1), 8-14 (2012-10-30)
Most chemotherapy regimens in colorectal cancer treatment are 5-fluorouracil (5-FU)/leucovorin or capecitabine-based. Cardiotoxicity is a less common but potentially lethal complication of 5-FU or capecitabine treatment, and some physicians might be unfamiliar with treatment alternatives. Rechallenging should be avoided because
John M L Ebos et al.
EMBO molecular medicine, 6(12), 1561-1576 (2014-11-02)
Thousands of cancer patients are currently in clinical trials evaluating antiangiogenic therapy in the neoadjuvant setting, which is the treatment of localized primary tumors prior to surgical intervention. The rationale is that shrinking a tumor will improve surgical outcomes and
Sharlene Gill et al.
Cancer treatment reviews, 40(10), 1171-1181 (2014-12-03)
Colorectal cancer (CRC) is the third most commonly diagnosed cancer among males and second among females worldwide. The treatment landscape for advanced CRC (aCRC) is rapidly evolving and there are now a number of randomized trials assessing treatment of aCRC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service