Merck
  • Laser diode thermal desorption atmospheric pressure chemical ionization tandem mass spectrometry applied for the ultra-fast quantitative analysis of BKM120 in human plasma.
Laser diode thermal desorption atmospheric pressure chemical ionization tandem mass spectrometry applied for the ultra-fast quantitative analysis of BKM120 in human plasma.
Analytical and bioanalytical chemistry (2014-06-25)
Christian Lanshoeft, Olivier Heudi, Luc Alexis Leuthold, Götz Schlotterbeck, Walid Elbast, Franck Picard, Olivier Kretz
ABSTRACT
A sensitive and ultra-fast method utilizing the laser diode thermal desorption ion source using atmospheric pressure chemical ionization coupled to tandem mass spectrometry (LDTD-APCI-MS/MS) was developed for the quantitative analysis of BKM120, an investigational anticancer drug in human plasma. Samples originating from protein precipitation (PP) followed by salting-out assisted liquid-liquid extraction (SALLE) were spotted onto the LazWell™ plate prior to their thermal desorption and detection by tandem mass spectrometry in positive mode. The validated method described in this paper presents a high absolute extraction recovery (>90 %) for BKM120 and its internal standard (ISTD) [D8]BKM120, with precision and accuracy meeting the acceptance criteria. Standard curves were linear over the range of 5.00 to 2000 ng mL(-1) with a coefficient of determination (R (2)) >0.995. The method specificity was demonstrated in six different batches of human plasma. Intra- and inter-run precision as well as accuracy within ±20 % at the lower limit of quantification (LLOQ) and ±15 % (other levels) were achieved during a three-run validation for quality control (QC) samples. The post-preparative stability on the LazWell™ plate at room temperature was 72 h and a 200-fold dilution of spiked samples was demonstrated. The method was applied successfully to three clinical studies (n = 847) and cross-checked with the validated LC-ESI-MS/MS reference method. The sample analysis run time was 10 s as compared to 4.5 min for the current validated LC-ESI-MS/MS method. The resultant data were in agreement with the results obtained using the validated reference LC-ESI-MS/MS assay and the same pharmacokinetic (PK) parameters were calculated for both analytical assays. This work demonstrates that LDTD-APCI-MS/MS is a reliable method for the ultra-fast quantitative analysis of BKM120 which can be used to speed-up and support its bioanalysis in the frame of the clinical trials.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material