Merck
  • Robust and selective nano cavities for protein separation: an interpenetrating polymer network modified hierarchically protein imprinted hydrogel.
Robust and selective nano cavities for protein separation: an interpenetrating polymer network modified hierarchically protein imprinted hydrogel.
Journal of chromatography. A (2014-05-03)
Ali Nematollahzadeh, Patrick Lindemann, Wei Sun, Jörg Stute, Dirk Lütkemeyer, Börje Sellergren
ABSTRACT
In the present work, we report a novel method for the reinforcement of hierarchically structured molecularly imprinted polymer (MIP) for the separation of human serum albumin (HSA) and immunoglobulin G (IgG) proteins under pressure driven flow conditions. The template proteins (HSA or IgG) were first physically adsorbed at their isoelectric point on the surface of wide pore silica particles. Thereafter, the pore system was filled with a monomer solution and polymerized to form a lightly crosslinked polyacrylamide network covering the protein template. In order to enhance the rigidity of the hydrogels, different type of crosslinkers such as ethylene glycol dimethacrylate (EGDMA), 3,4-dihydroxyphenethylamine (DA) and methylene-bis-acrylamide (MBA), at least partially interpenetrated into the initially made acrylamide base hydrogel leading to formation of an interpenetrating polymer network (IPN). Then the silica matrix was removed to leave highly porous and reinforced MIP. TGA together with FT-IR and TEM analysis supported the interpenetration of the secondary crosslinkers in the initially formed polymer matrix. The compression property of the modified hydrogels as a function of degree of swelling was in the following order: DA>EGDMA>MBA-IPN-modified hydrogel. Batch binding assay verified the capability of the IPN modified MIPs to capture the target proteins. Moreover, solid phase extraction, HPLC and SDS-PAGE revealed that the EGDMA modified MIP could selectively capture and separate the proteins from human serum or fermentation broth.
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Methylamine, ≥99.0%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis