Merck
  • Environmental fate of the next generation refrigerant 2,3,3,3-tetrafluoropropene (HFO-1234yf).
Environmental fate of the next generation refrigerant 2,3,3,3-tetrafluoropropene (HFO-1234yf).
Environmental science & technology (2014-10-21)
Jeongdae Im, Gillian E Walshe-Langford, Ji-Won Moon, Frank E Löffler
ABSTRACT
The hydrofluoroolefin 2,3,3,3-tetrafluoropropene (HFO-1234yf) has been introduced to replace 1,1,1,2-tetrafluoroethane (HFC-134a) as refrigerant in mobile, including vehicle, air conditioning systems because of its lower global warming potential. HFO-1234yf is volatile at ambient temperatures; however, high production volumes and widespread handling are expected to release this fluorocarbon into terrestrial and aquatic environments, including groundwater. Laboratory experiments explored HFO-1234yf degradation by (i) microbial processes under oxic and anoxic conditions, (ii) abiotic processes mediated by reactive mineral phases and zerovalent iron (Fe(0), ZVI), and (iii) cobalamin-catalyzed biomimetic transformation. These investigations demonstrated that HFO-1234yf was recalcitrant to microbial (co)metabolism and no transformation was observed in incubations with ZVI, makinawite (FeS), sulfate green rust (GR(SO4)), magnetite (Fe(3)O(4)), and manganese oxide (MnO2). Sequential reductive defluorination of HFO-1234yf to 3,3,3-trifluoropropene and 3,3-dichloropropene with concomitant stoichiometric release of fluoride occurred in incubations with reduced cobalamins (e.g., vitamin B12) indicating that biomolecules can transform HFO-1234yf at circumneutral pH and at ambient temperature. Taken together, these findings suggest that HFO-1234yf recalcitrance in aquifers should be expected; however, HFO-1234yf is not inert and a biomolecule may mediate reductive transformation in low redox environments, albeit at low rates.
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Supelco
Tetrachloroethylene, analytical standard
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Tetrachloroethylene, anhydrous, ≥99%
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Sigma-Aldrich
Methane, electronic grade, ≥99.998%
Sigma-Aldrich
Tetrachloroethylene, ACS reagent, ≥99.0%
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Trichloroethylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Tetrachloroethylene, ≥99.5%
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Vitamin B12, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Tetrachloroethylene, suitable for HPLC, ≥99.9%
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Density Standard 1623 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod