Merck
  • Mesoporous titania thin films as efficient enzyme carriers for paraoxon determination/detoxification: effects of enzyme binding and pore hierarchy on the biocatalyst activity and reusability.
Mesoporous titania thin films as efficient enzyme carriers for paraoxon determination/detoxification: effects of enzyme binding and pore hierarchy on the biocatalyst activity and reusability.
The Analyst (2014-05-03)
N Frančič, M G Bellino, G J A A Soler-Illia, A Lobnik
ABSTRACT
In this work we demonstrate the efficient immobilization of histidine 6-tagged organophosphate hydrolase (His6-OPH), an organophosphate-degrading enzyme, on mesoporous titania thin films. This permits the use of the biocatalyst films as efficient tools in the detection/detoxification of paraoxon. His6-OPH was immobilized on mesoporous thin films with uniform (9 nm) and bimodal (13-38 nm) pore size distribution, through covalent attachment and physical adsorption. The biocatalyst films show good activity, and enhanced stability with respect to the free enzyme at extreme conditions of pH and temperature, especially around neutral pH and room temperature. In addition, the bioactive films can be easily separated from the reaction media and reused multiple times without significant loss of activity.
Product Description
Supelco
Tetrahydrofuran, analytical standard
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, suitable for plant cell culture
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, reagent grade
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
SAFC
HEPES
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., 98-102%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, ACS reagent, 98%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%