Skip to Content
Merck
All Photos(1)

Key Documents

87369

Supelco

Tetrahydrofuran

≥99.5%, ≥99.5% (GC), for ion-selective electrodes, contains 250 ppm BHT as inhibitor, Selectophore

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$115.00
500 ML
$348.00

$115.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 ML
$115.00
500 ML
$348.00

About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NB.61
grade:
for ion-selective electrodes
Assay:
≥99.5% (GC)
≥99.5%
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

$115.00


In StockDetails


Request a Bulk Order

Product Name

Tetrahydrofuran, Selectophore, ≥99.5%

grade

for ion-selective electrodes

Quality Level

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

description

solvent for preparation of membranes

product line

Selectophore

Assay

≥99.5% (GC)
≥99.5%

form

liquid

autoignition temp.

610 °F

contains

250 ppm BHT as inhibitor

expl. lim.

1.8-11.8 %

impurities

≤0.01% water

refractive index

n20/D 1.407 (lit.)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

solubility

H2O: soluble

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran (THF), an organic compound which belongs to saturated cyclic ether group. It is mainly used as solvent for PVC and organoalkali compounds. It is also used in lithium batteries.
Visit our Sensor Applications portal to learn more.

Application

THF has been used to realise PVC membrane from electrode, during a study to design, fabricate and electrochemically characterize miniaturised thin- and thick-film Ag/AgCl reference electrodes.[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

recommended

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strategies of miniaturised reference electrodes integrated in a silicon based ?one chip? pH sensor.
Simonis, Anette, et al.
Sensors, 3.9, 330-339 (2003)
Luca Chiari et al.
The Journal of chemical physics, 138(7), 074301-074301 (2013-03-01)
We present total, elastic, and inelastic cross sections for positron and electron scattering from tetrahydrofuran (THF) in the energy range between 1 and 5000 eV. Total cross sections (TCS), positronium formation cross sections, the summed inelastic integral cross sections (ICS)
C N Ong et al.
British journal of industrial medicine, 48(9), 616-621 (1991-09-01)
Occupational exposure to tetrahydrofuran (THF) was studied by analysis of environmental air, blood, alveolar air, and urine from 58 workers in a video tape manufacturing plant. Head space gas chromatography (GC) with an FID detector was used for determination of
Kenneth F Ilett et al.
Drug metabolism and disposition: the biological fate of chemicals, 30(9), 1005-1012 (2002-08-09)
The aim of this study was to elucidate the metabolic pathways for dihydroartemisinin (DHA), the active metabolite of the artemisinin derivative artesunate (ARTS). Urine was collected from 17 Vietnamese adults with falciparum malaria who had received 120 mg of ARTS
Eliane M de Oliveira et al.
Physical chemistry chemical physics : PCCP, 15(5), 1682-1689 (2012-12-19)
We report elastic integral, differential and momentum transfer cross sections for low-energy electron scattering by the cellulose components β-D-glucose and cellobiose (β(1 → 4) linked glucose dimer), and the hemicellulose component β-D-xylose. For comparison with the β forms, we also

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service