Merck
  • Environmentally relevant concentrations of galaxolide (HHCB) and tonalide (AHTN) induced oxidative and genetic damage in Dreissena polymorpha.
Environmentally relevant concentrations of galaxolide (HHCB) and tonalide (AHTN) induced oxidative and genetic damage in Dreissena polymorpha.
Journal of hazardous materials (2014-12-03)
Marco Parolini, Stefano Magni, Irene Traversi, Sara Villa, Antonio Finizio, Andrea Binelli
ABSTRACT
Synthetic musk compounds (SMCs) are extensively used as fragrances in several personal care products and have been recognized as emerging aquatic pollutants. Among SMCs, galaxolide (HHCB) and tonalide (AHTN) are extensively used and have been measured in aquatic ecosystems worldwide. However, their potential risk to organisms remains largely unknown. The aim of this study was to investigate whether 21-day exposures to HHCB and AHTN concentrations frequently measured in aquatic ecosystems can induce oxidative and genetic damage in Dreissena polymorpha. The lipid peroxidation (LPO) and protein carbonyl content (PCC) were measured as oxidative stress indexes, while the DNA precipitation assay and the micronucleus test (MN test) were applied to investigate genetic injuries. HHCB induced significant increases in LPO and PCC levels, while AHTN enhanced only protein carbonylation. Moreover, significant increases in DNA strand breaks were caused by exposure to the highest concentrations of HHCB and AHTN tested in the present study, but no fixed genetic damage was observed.
Product Description
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer