Merck
  • Upregulated plasma and urinary levels of nucleosides as biological markers in the diagnosis of primary gallbladder cancer.
Upregulated plasma and urinary levels of nucleosides as biological markers in the diagnosis of primary gallbladder cancer.
Journal of separation science (2014-08-20)
Xingyuan Jiao, Yuxuan Mo, Ying Wu, Jinyun He, Peng Zhang, Ronglin Hu, Canqiao Luo, Jun Du, Jian Fu, Jinsen Shi, Liansuo Zhou, Dongming Li
ABSTRACT
We first detected aberrant nucleoside levels in the plasma, urine, bile, and tissues from cases and controls to explore them as biomarkers in the diagnosis of gallbladder cancer. Reversed-phase high-performance liquid chromatography was used to assess the levels of ten nucleosides in these samples from gallbladder cancer patients, gallstone patients, and healthy controls. Plasma and urine samples were collected from patients with gallbladder cancer (n = 202), patients with gallstones (n = 203), and healthy controls (n = 205); bile and tissue samples were collected from 91 gallbladder cancer patients, 93 gallstone patients; and 90 were donated after cardiac death. Of the ten nucleosides analyzed, eight urinary nucleosides, five plasma nucleosides, three bile nucleosides, and one tissue nucleoside were significantly upregulated in the gallbladder cancer patients compared to control groups (p < 0.05). Among these upregulated nucleosides, the sensitivity, specificity, and accuracy of urinary nucleosides in the diagnosis of gallbladder cancer patients were 89.4, 97.1, and 95.7%, respectively, those of plasma nucleosides were 91.2, 95.6, and 94.2%, respectively, those of bile nucleosides were 95.3, 96.4, and 95.1%, respectively, and those of tissue nucleosides were 86.2, 93.8, and 92.6%, respectively. These results suggest that nucleosides may be as useful as biological markers for gallbladder cancer.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%