Merck
  • Development of the microsporidian parasite, Loma salmonae, in a rainbow trout gill epithelial cell line (RTG-1): evidence of xenoma development in vitro.
Development of the microsporidian parasite, Loma salmonae, in a rainbow trout gill epithelial cell line (RTG-1): evidence of xenoma development in vitro.
Parasitology (2014-12-02)
S H McConnachie, J Sheppard, G M Wright, D J Speare
ABSTRACT
Growth and propagation of fish-infecting microsporidians within cell culture has been more difficult to achieve than for insect- and human-infecting microsporidians. Fish microsporidia tend to elicit xenoma development rather than diffuse growth in vivo, and this process likely increases host specificity. We present evidence that the fish microsporidian, Loma salmonae, has the capacity to develop xenomas within a rainbow trout gill epithelial cell line (RTG-1). Spore numbers increased over a 4 weeks period within cell culture flasks. Xenoma-like structures were observed using phase contrast microscopy, and then confirmed using transmission electron microscopy. Optimization of the L. salmonae-RTG-1 cell model has important implications in elucidating the process of xenoma development induced by microsporidian parasites.
Product Description
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium tetraborate, 99%
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY))
SAFC
L-Glutamine
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
USP
Benzocaine, United States Pharmacopeia (USP) Reference Standard
Benzocaine, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)