Merck
  • Poly(L-lactide) and poly(butylene succinate) immiscible blends: from electrospinning to biologically active materials.
Poly(L-lactide) and poly(butylene succinate) immiscible blends: from electrospinning to biologically active materials.
Materials science & engineering. C, Materials for biological applications (2014-06-09)
Nikoleta Stoyanova, Dilyana Paneva, Rosica Mincheva, Antoniya Toncheva, Nevena Manolova, Philippe Dubois, Iliya Rashkov
ABSTRACT
For the first time the preparation of defect-free fibers from immiscible blends of high molar mass poly(lactic acid) (PLA) and poly(butylene succinate) (PBS) in the whole range of the polyester weight ratios is shown. Electrospinning using the solvent-nonsolvent approach proved most appropriate. Moreover, electrospinning revealed crucial for the obtaining of PLA/PBS materials maintaining integrity. DSC and XRD analyses attested for a plasticizing effect and for increased PLA crystallinity at PBS addition to PLA. The mechanical properties of the PLA/PBS mats were controlled by the alignment of the fibers and changed from plastic to brittle materials upon increasing the PBS content. Drug loading and tests against pathogenic microorganisms suggested that the obtained mats can find application as antibacterial fibrous materials.
Product Description
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
5-Chloro-8-quinolinol, 95%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer