Merck
  • Solid lipid nanoparticles of anticancer drug andrographolide: formulation, in vitro and in vivo studies.
Solid lipid nanoparticles of anticancer drug andrographolide: formulation, in vitro and in vivo studies.
Drug development and industrial pharmacy (2013-07-06)
Rabea Parveen, F J Ahmad, Z Iqbal, M Samim, Sayeed Ahmad
ABSTRACT
Diterpenoidal anti-cancer drug andrographolide (AD) was encapsulated into solid lipid nanoparticle (SLN) because of poor aqueous solubility and high lipophilicity. AD-SLNs were prepared by solvent injection method and characterized for droplet size, surface morphology, zeta potential, etc. In vitro drug release was carried out by dialysis-membrane method. A pharmacokinetic study was performed by UPLC/Q-TOF-MS method to determine the maximum plasma concentration (Cmax), area under the curve (AUC), etc. There was an improvement in Cmax and AUC of AD-SLNs when compared with AD, thereby enhancing the bioavailability of AD. The tmax was increased than that of AD suspension, indicating the sustained release pattern of AD-SLNs. The antitumor activity was carried out on Balb/c mice showing better results with AD-SLNs as compared to AD. Thus, the AD-loaded SLNs would be useful for delivering poorly water-soluble AD with enhanced bioavailability and improved antitumor activity.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles