Merck
  • Isolation of antifungal compound from Paeonia suffruticosa and its antifungal mechanism.
Isolation of antifungal compound from Paeonia suffruticosa and its antifungal mechanism.
Chinese journal of integrative medicine (2014-03-01)
Yong Zhao, Bao-en Wang, Shu-wen Zhang, Shu-min Yang, Hong Wang, Ai-min Ren, En-tong Yi
ABSTRACT
To isolate antifungal compound from Paeonia suffruticosa, and to find the antifungal mechanisms by observing the ultrastructural modifications of yeasts in growth phase produced by 1,2,3,4,6-penta-O-galloyl-beta-D-glucose (PGG). Peony (Paeonia suffruticosa) root bark (PRB) was separated by solvent extraction and purified by high performance liquid chromatography (HPLC) method using analytical and preparative reversed phase C18 column on the basis of bio-assay method. In order to investigate the antifungal mechanism of PGG, Yeasts were submitted to different concentrations [3 × minimum inhibition concentration (MIC), 0.3 × MIC] for 1 h under constant stirring at 30 °C, and transmission electron microscopy was performed. Based on the antifungal activity of PRB on Candida glabrata CBS138, the antifungal compound were isolated in ethyl acetate layer of PRB and identified as PGG by mass spectrometry, 1H nuclear magnetic resonance (NMR) analyses, with molecular weight of 940 and molecular formular as C41H32O26. Transmission electron microscopy showed that PGG degraded the cell wall envelope. The results suggest that PGG may be responsible for the antifungal activity of PRB by disrupting the structure of cell wall directly.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Penta-O-galloyl-β-D-glucose hydrate, ≥96% (HPLC)
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%