Merck
  • Molecular imprinted polymeric porous layers in open tubular capillaries for chiral separations.
Molecular imprinted polymeric porous layers in open tubular capillaries for chiral separations.
Journal of chromatography. A (2014-06-18)
Chadin Kulsing, Radim Knob, Mirek Macka, Paul Junor, Reinhard I Boysen, Milton T W Hearn
ABSTRACT
A new method has been developed for the preparation of molecular imprinted polymers as porous layers in open tubular (MIP-PLOT) capillary column formats for use in chiral separations by capillary liquid chromatography. The synthesis was based on 'in-capillary' ultraviolet (UV) initiated polymerization using light emitting diodes (LEDs) in conjunction with the continuous delivery of the pre-polymerization reagents into the polymerization zone of the capillary using an automated capillary delivery device. The relationships between exposure times, UV-light intensity and polymer layer thickness have been determined, as well as the effects of reagent delivery rate and multiple LED exposures on the layer thickness for various compositions of pre-polymerization mixtures. The polymer surface morphology was investigated by scanning electron microscopy (SEM). The non-steroidal anti-inflammatory drug S-ketoprofen was used as the template for the preparation of the MIP imprinted PLOT coatings. The separation performance with the ketoprofen racemate was investigated by capillary liquid chromatography. In contrast to alternative methods, which require the use of expensive chiral selectors, the described MIP PLOT stationary phases used non-chiral polymer precursors to create enantioselective nano-cavities through molecular self-assembly processes. The described fabrication methods provide a new avenue to tailor-make chiral MIP-PLOT capillary columns for the separation of chiral compounds present in complex or racemic analyte mixtures of chemical and biological origin.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%