Merck
  • Synthesis and evaluation of the novel 2-[¹⁸F]fluoro-3-propoxy-triazole-pyridine-substituted losartan for imaging AT₁ receptors.
Synthesis and evaluation of the novel 2-[¹⁸F]fluoro-3-propoxy-triazole-pyridine-substituted losartan for imaging AT₁ receptors.
Bioorganic & medicinal chemistry (2014-07-16)
Natasha Arksey, Tayebeh Hadizad, Basma Ismail, Maryam Hachem, Ana C Valdivia, Rob S Beanlands, Robert A deKemp, Jean N DaSilva
ABSTRACT
The 2-[(18)F]fluoro-3-pent-4-yn-1-yloxypyridine ([(18)F]FPyKYNE) analog of the potent non-peptide angiotensin II type 1 receptor (AT1R) blocker losartan was produced via click chemistry linking [(18)F]FPyKYNE to azide-modified tetrazole-protected losartan followed by TFA deprotection. Preliminary small animal imaging with positron emission tomography (PET) in rats displayed high uptake in the kidneys with good contrast to surrounding tissue. Rat metabolism displayed the presence of 23% unchanged tracer in plasma at 30 min. Upon co-administration with AT₁R blocker candesartan (2.5, 5 and 10 mg/kg), a dose-dependent reduction (47-65%) in tracer uptake was observed in the kidney, while no difference was observed following AT₂R blocker PD123,319 (5 mg/kg), indicating binding selectivity for AT₁R over AT₂R and potential for imaging AT₁R using PET.
Product Description
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Supelco
Losartan potassium
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Losartan potassium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
USP
Losartan potassium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Diethyl ether, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Pentane, ≥99% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Pentane, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Pentane, puriss. p.a., ≥99.0% (GC)