Merck
  • Inhibition of beta cell growth and function by bone morphogenetic proteins.
Inhibition of beta cell growth and function by bone morphogenetic proteins.
Diabetologia (2014-09-28)
Christine Bruun, Gitte L Christensen, Marie L B Jacobsen, Marianne B Kanstrup, Pernille R Jensen, Helle Fjordvang, Thomas Mandrup-Poulsen, Nils Billestrup
ABSTRACT
Impairment of beta cell mass and function is evident in both type 1 and type 2 diabetes. In healthy physiological conditions pancreatic beta cells adapt to the body's increasing insulin requirements by proliferation and improved function. We hypothesised that during the development of diabetes, there is an increase in the expression of inhibitory factors that prevent the beta cells from adapting to the increased need for insulin. We evaluated the effects of bone morphogenetic protein (BMP) 2 and -4 on beta cells. The effects of BMP2 and -4 on beta cell proliferation, apoptosis, gene expression and insulin release were studied in isolated islets of Langerhans from rats, mice and humans. The expression of BMPs was analysed by immunocytochemistry and real-time PCR. The role of endogenous BMP was investigated using a soluble and neutralising form of the BMP receptor 1A. BMP2 and -4 were found to inhibit basal as well as growth factor-stimulated proliferation of primary beta cells from rats and mice. Bmp2 and Bmp4 mRNA and protein were expressed in islets and regulated by inflammatory cytokines. Neutralisation of endogenous BMP activity resulted in enhanced proliferation of rodent beta cells. The expression of Id mRNAs was induced by BMP4 in rat and human islets. Finally, glucose-induced insulin secretion was significantly impaired in rodent and human islets pre-treated with BMP4, and inhibition of BMP activity resulted in enhanced insulin release. These data show that BMP2 and -4 exert inhibitory actions on beta cells in vitro and suggest that BMPs exert regulatory roles of beta cell growth and function.
Sigma-Aldrich
Anti-SMAD2 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%