Skip to Content
Merck
All Photos(4)

Documents

AV38694

Sigma-Aldrich

Anti-SMAD2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-SMAD family member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

mouse, sheep, guinea pig, rabbit, human, dog, bovine, horse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SMAD2(4087)

Immunogen

Synthetic peptide directed towards the N terminal region of human SMAD2

Biochem/physiol Actions

SMAD proteins are transcription regulators that mediate multiple signal transduction pathways. SMAD2 mediates the TGF-β pathway that is involved in a wide variety of cellular processes such as cell growth, apoptosis, survival, differentiation, morphogenesis and immune responses. SMAD/TGF-β signaling accelerates skin wound healing.

Sequence

Synthetic peptide located within the following region: VVKRLLGWKKSAGGSGGAGGGEQNGQEEKWCEKAVKSLVKKLKKTGRLDE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Kyoung Jun et al.
International journal of molecular sciences, 15(1), 605-628 (2014-01-09)
In a previous study, we isolated human amniotic fluid (AF)-derived mesenchymal stem cells (AF-MSCs) and utilized normoxic conditioned medium (AF-MSC-norCM) which has been shown to accelerate cutaneous wound healing. Because hypoxia enhances the wound healing function of mesenchymal stem cell-conditioned
Toshio Matsumoto et al.
Endocrine journal, 59(2), 91-101 (2011-09-21)
Mechanical stress and parathyroid hormone (PTH) are major stimulators, and aging and glucocorticoids excess are important suppressors of osteoblast differentiation. Mechanical stress and PTH stimulate interleukin (IL)-11 expression in cells of osteoblast lineage by enhancing transcription of IL-11 gene via
Yi Yang et al.
Oncology research, 21(6), 345-352 (2013-01-01)
TGF-β/Smad signaling induces epithelial-mesenchymal transition (EMT) and tumor metastasis. As essential mediators in this pathway, Smad2 and Smad3 have been extensively studied and found to promote EMT and the subsequent mobility as well as invasiveness of lung cancer cells. In
Hye Sook Min et al.
Laboratory investigation; a journal of technical methods and pathology, 94(6), 598-607 (2014-04-02)
Dipeptidyl peptidase IV (DPPIV) is an exopeptidase that modulates the function of several substrates, among which insulin-releasing incretin hormones are the most well known. DPPIV also modulate substrates involved in inflammation, cell migration, and cell differentiation. Although DPPIV is highly
Minghua Wu et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(4), 1010-1021 (2014-04-24)
Systemic sclerosis (SSc) is a chronic autoimmune disease clinically manifesting as progressive fibrosis of the skin and internal organs. Recent microarray studies demonstrated that cadherin 11 (Cad-11) expression is increased in the affected skin of patients with SSc. The purpose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service