Merck
  • Folate-conjugated beta-cyclodextrin-based polymeric micelles with enhanced doxorubicin antitumor efficacy.
Folate-conjugated beta-cyclodextrin-based polymeric micelles with enhanced doxorubicin antitumor efficacy.
Colloids and surfaces. B, Biointerfaces (2014-07-25)
Lu Zhang, Jiafei Lu, Yangmin Jin, Liyan Qiu
ABSTRACT
In order to enhance the antitumor effects of doxorubicin (DOX), a novel micellar vector with high DOX loading and tumor targeting function based on folate-conjugated amphiphilic copolymer folate-poly(ethylene glycol)-poly(d,l-lactide)-β-cyclodextrin (FA-PEL-CD) was constructed. Cytotoxicity and cellular uptake experiments were performed in HeLa, KB, and A549 cell lines expressing different amounts of folate receptors in order to evaluate the targeting effect of the folate modification. The antitumor experiments performed in a KB cell-xenografted nude mouse model showed that the treatment with 10mg/kg DOX loaded FA-PEL-CD micelles achieved approximately 86% of tumor growth inhibition compared to the control. Ex vivo fluorescence imaging experiments and histological examination confirmed that folate modification can enhance the antitumorigenesis efficacy and reduce the cardiotoxicity of DOX. These results suggest that FA-PEL-CD copolymer-based micelles are promising nanocarriers for targeted doxorubicin delivery, with improved antitumor efficacy and reduced toxicity in normal tissues.
Product Description
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Supelco
Toluene, analytical standard
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol