Skip to Content
Merck
All Photos(1)

Key Documents

W424601

Sigma-Aldrich

Triethylamine

≥99.5%

Synonym(s):

N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing

Select a Size

W424601-SAMPLE
$83.00
1 KG
$154.00
20 KG
$831.00

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W424601-SAMPLE
$83.00
1 KG
$154.00
20 KG
$831.00

About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
FEMA Number:
4246
Beilstein:
605283
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Flavis number:
11.023
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

reg. compliance

FDA 21 CFR 117

vapor density

3.5 (vs air)

vapor pressure

51.75 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

593 °F

shelf life

5 yr

expl. lim.

8 %

refractive index

n20/D 1.401 (lit.)

pH

12.7 (15 °C, 100 g/L)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylamine may be used as a test volatile organic compound (VOC) in studies of the development of sensors for detecting VOCs.[1]

Application


  • Novel hybrid thiazoles, bis-thiazoles linked to azo-sulfamethoxazole: Synthesis, docking, and antimicrobial activity.: This study showcases the synthesis of hybrid thiazoles linked to azo-sulfamethoxazole, employing triethylamine in the reaction process, highlighting its role in antimicrobial applications (Salem et al., 2024).

  • Spirocyclic rhodamine B benzoisothiazole derivative: a multi-stimuli fluorescent switch manifesting ethanol-responsiveness, photo responsiveness, and acidochromism.: This research demonstrates a spirocyclic rhodamine derivative acting as a multi-stimuli-responsive fluorescent switch, where triethylamine plays a critical role in the synthesis process (Battula et al., 2023).

  • Para-Substituted Thiosemicarbazones as Cholinesterase Inhibitors: Synthesis, In Vitro Biological Evaluation, and In Silico Study.: This paper reports on the synthesis of para-substituted thiosemicarbazones where triethylamine is employed as a base, evaluating their potential as cholinesterase inhibitors (Khan et al., 2023).

  • Separation and purification of quinolyridine alkaloids from seeds of Thermopsis lanceolata R. Br. by conventional and pH-zone-refining counter-current chromatography.: This study involves the use of triethylamine in the pH-zone-refining counter-current chromatography technique to separate and purify specific alkaloids, illustrating its utility in advanced separation methodologies (Ning et al., 2023).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porphyrins-based opto-electronic nose for volatile compounds detection.
Di Natale C, et al.
Sensors and Actuators B, Chemical, 65(1), 220-226 (2000)
Kai Morino et al.
Scientific reports, 5, 8953-8953 (2015-05-21)
Well-trained clinicians may be able to provide diagnosis and prognosis from very short biomarker series using information and experience gained from previous patients. Although mathematical methods can potentially help clinicians to predict the progression of diseases, there is no method
Sumanta Kumar Padhi et al.
Inorganic chemistry, 51(15), 8091-8102 (2012-07-26)
Cyclometalated ruthenium complexes having C(^)N and N(^)C type coordinating ligands with NAD(+)/NADH function have been synthesized and characterized by spectroscopic methods. The variation of the coordinating position of σ-donating carbon atom leads to a drastic change in their properties. Both
Hannah Coyle et al.
The American journal of geriatric psychiatry : official journal of the American Association for Geriatric Psychiatry, 23(4), 335-359 (2014-07-08)
The aim of this study was to assess the efficacy of cognitive training, specifically computerized cognitive training (CCT) and virtual reality cognitive training (VRCT), programs for individuals living with mild cognitive impairment (MCI) or dementia and therefore at high risk
André R Fajardo et al.
Biomacromolecules, 15(7), 2691-2700 (2014-05-27)
Despite a growing interest in amphiphilic polysaccharide-based diblock copolymers as functional polymeric drug delivery nanosystems, biologically relevant sulfated glycosaminoglycan systems were not yet investigated. Here, we report the synthesis and the self-assembly properties in water of chondroitin sulfate-b-poly(lactic acid) (CS-b-PLA(n)).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service