Merck
  • Quantitation of the immunological adjuvants, monophosphoryl lipid A and Quil A in poly (lactic-co-glycolic acid) nanoparticles using high performance liquid chromatography with evaporative light scattering detection.
Quantitation of the immunological adjuvants, monophosphoryl lipid A and Quil A in poly (lactic-co-glycolic acid) nanoparticles using high performance liquid chromatography with evaporative light scattering detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-02)
Sharan Bobbala, Arlene McDowell, Sarah Hook
ABSTRACT
Monophosphoryl lipid A (MPL) and Quil A are two immunological adjuvants commonly used in vaccines. At present no simple, validated methods for the quantification of Quil A and MPL have been previously reported therefore the aim of the current study was to develop a simple, fast and validated method to quantify MPL and Quil A using high performance liquid chromatography evaporative light scattering detection (HPLC-ELSD). The HPLC-ELSD technique was carried out using a ZORBAX Eclipse XDB-C8 column (2.1×50 mm; particle size, 3.5 μm) in an isocratic elution mode at 25 °C. MPL was eluted at a retention time of 1.8 min with methanol-water as the mobile phase and a detector temperature of 75 °C. Quil A was resolved as three peaks with retention times of 4.1, 5.5 and 6.4 min with a detector temperature of 30 °C and with water-acetonitrile and 0.01% formic acid as the mobile phase. The nebulizer pressure and gain were set at 3.5 bar and 10, respectively. Calibration curves plotted for both the adjuvants had an R(2)>0.997. Accuracy, intra- and inter-day precision were within the accepted limits. The limit of detection for MPL and Quil A were calculated as 1.343 and 2.06 μg/mL, respectively. The limit of quantification was 2.445 for MPL and 8.97 μg/mL for Quil A. This analytical method was used to quantify the entrapment and in vitro release of MPL and Quil A in a poly lactic-co-glycolic acid (PLGA) nanoparticle vaccine.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Supelco
Methanol, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer