Merck
  • Development of a protective dermal drug delivery system for therapeutic DNAzymes.
Development of a protective dermal drug delivery system for therapeutic DNAzymes.
International journal of pharmaceutics (2014-12-30)
Kay Marquardt, Anna-Carola Eicher, Dorota Dobler, Ulf Mäder, Thomas Schmidts, Harald Renz, Frank Runkel
ABSTRACT
RNA-cleaving DNAzymes are a potential novel class of nucleic acid-based active pharmaceutical ingredients (API). However, developing an appropriate drug delivery system (DDS) that achieves high bioavailability is challenging. Especially in a dermal application, DNAzymes have to overcome physiological barriers composed of penetration barriers and degrading enzymes. The focus of the present study was the development of a protective and penetration-enhanced dermal DDS that was tailor made for DNAzymes. DNAzyme Dz13 was used as a potential API for topical therapy against actinic keratosis. In the progress of development and selection, different preservatives, submicron emulsions (SMEs) and the physiological pH range were validated with respect to the API's integrity. A physicochemical stable SME of a pharmaceutical grade along with a high API integrity was achieved. Additionally, two developed protective systems, consisting of a liposomal formulation or chitosan-polyplexes, reduced the degradation of Dz13 in vitro. A combination of SME and polyplexes was finally validated at the skin and cellular level by in vitro model systems. Properties of penetration, degradation and distribution were determined. The result was enhanced skin penetration efficiency and increased cellular uptake with a high protective efficiency for DNAzymes due to the developed protective DDS.
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Stearyl alcohol, European Pharmacopoeia (EP) Reference Standard
USP
Stearyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%