Merck
  • Liposomal encapsulation of dexamethasone modulates cytotoxicity, inflammatory cytokine response, and migratory properties of primary human macrophages.
Liposomal encapsulation of dexamethasone modulates cytotoxicity, inflammatory cytokine response, and migratory properties of primary human macrophages.
Nanomedicine : nanotechnology, biology, and medicine (2014-03-13)
Matthias Bartneck, Franziska Marie Peters, Klaudia Theresa Warzecha, Michaela Bienert, Louis van Bloois, Christian Trautwein, Twan Lammers, Frank Tacke
ABSTRACT
The encapsulation of drugs into liposomes aims to enhance their efficacy and reduce their toxicity. Corticosteroid-loaded liposomes are currently being evaluated in patients suffering from rheumatoid arthritis, atherosclerosis, colitis, and cancer. Here, using several different fluorophore-labeled formulations, we comprehensively studied the impact of liposome encapsulation of the prototypic corticosteroid dexamethasone on various primary human cells in vitro. Liposomal dexamethasone targeted several primary cell types in a dose and time-dependent manner, but specifically reduced cytotoxicity against human fibroblasts and macrophages in comparison to the solute drug. Furthermore, macrophage maturation and polarization markers were altered. Interestingly, liposomal dexamethasone induced proinflammatory cytokine secretion (specifically TNF, IL1β, IL6) in unstimulated cells, but reduced this response under inflammatory conditions. Monocyte and macrophage migration was significantly inhibited by dexamethasone-loaded liposomes. The findings indicate that the encapsulation of dexamethasone into liposomes modulates their cellular mechanism of action, and provides important indications for follow-up in vivo investigations. This study investigates mechanism of action of liposomal dexamethason in the treatment of inflammatory conditions. It is concluded that liposomal dexamethasone actually induces proinflammatory cytokine secretion in unstimulated cells, but reduces the same response under inflammatory conditions. Monocyte and macrophage migration was also inhibited. The findings indicate that liposomal dexamethasone may have different mechanisms of action than its native counterpart.
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Pyrrole, analytical standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Dexamethasone, VETRANAL®, analytical standard
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
MCP-1/MCAF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard