Merck
  • Stability-indicating chromatographic methods for the determination of sertindole.
Stability-indicating chromatographic methods for the determination of sertindole.
Journal of chromatographic science (2013-06-05)
Nariman A El-Ragehy, Nagiba Y Hassan, Mohamed Abdelkawy, Mahmoud A Tantawy
ABSTRACT
In this work, two chromatographic methods have been developed and validated for the determination of sertindole (an antipsychotic agent) in the presence of its oxidative degradation product. Sertindole was subjected to stress stability studies, including acid, alkali, oxidative, photolytic and thermal degradation. The chromatographic methods included the use of thin-layer chromatography (TLC-densitometry) and high-performance liquid chromatography (HPLC). The TLC method employed aluminum TLC plates precoated with silica gel G.F254 as the stationary phase and methanol-ethyl acetate-33% ammonia (1:9:0.1, by volume) as the mobile phase, and the chromatograms were scanned at 227 nm. The developed HPLC method used a reversed-phase C18 column with isocratic elution. The mobile phase was composed of phosphate buffer pH 3.0-acetonitrile-triethylamine (45:55:0.03, by volume) and run at a flow rate of 1.0 mL/min. Quantitation was achieved with ultraviolet detection at 256 nm. The linearity ranges were found to be 2-14 µg/band and 5-200 µg/mL for TLC and HPLC, respectively. The developed methods were validated according to the International Conference on Harmonization guidelines and were applied for bulk powder and dosage forms.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)