Merck
Molecular cytogenetics of human single pronucleated zygotes.
Reproductive sciences (Thousand Oaks, Calif.) (2014-04-11)
Ana Raquel Azevedo, Maria João Pinho, Joaquina Silva, Rosália Sá, Sólveig Thorsteinsdóttir, Alberto Barros, Mário Sousa
ABSTRACT
The aim of the present study was to use fluorescence in situ hybridization to analyze the chromosome status of zygotes with a single pronucleus from in vitro fertilization (IVF) and intracytoplasmic sperm injection (ICSI) treatment cycles. In addition, we performed immunocytochemical detection of nuclear lamins and histone H3 trimethylated at lysine-9, Me(3)H3K9. Zygotes were processed 24 hours after insemination or injection to assure the absence of asynchrony. In opposition to previous results, we observed 2 pronuclei in 16 of 18 IVF zygotes and 40 of 64 ICSI zygotes, suggesting premature pronuclear breakdown. In IVF and ICSI zygotes, the rate of normal diploidy was only 6 of 16 and 27 of 56, respectively, suggesting that monopronucleated zygotes should not be used in assisted reproductive treatments. The possible mechanisms are discussed and compared to previous studies of monopronucleated zygotes.
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Triton X-100, laboratory grade