Merck
  • A multiscale entropy-based tool for scoring severity of systemic inflammation.
A multiscale entropy-based tool for scoring severity of systemic inflammation.
Critical care medicine (2014-04-11)
Benjamin Vandendriessche, Harlinde Peperstraete, Elke Rogge, Peter Cauwels, Eric Hoste, Oliver Stiedl, Peter Brouckaert, Anje Cauwels
ABSTRACT
Early detection and start of appropriate treatment are highly correlated with survival of sepsis and septic shock, but the currently available predictive tools are not sensitive enough to identify patients at risk. Linear (time and frequency domain) and nonlinear (unifractal and multiscale complexity dynamics) measures of beat-to-beat interval variability were analyzed in two mouse models of inflammatory shock to determine if they are sensitive enough to predict outcome. University research laboratory. Blood pressure transmitter-implanted female C57BL/6J mice. IV administration of tumor necrosis factor (n = 11) or lipopolysaccharide (n = 14). Contrary to linear indices of variability, unifractal dynamics, and absolute heart rate or blood pressure, quantification of complex beat-to-beat dynamics using multiscale entropy was able to predict survival outcome starting as early as 40 minutes after induction of inflammatory shock. Based on these results, a new and clinically relevant index of multiscale entropy was developed that scores the key features of a multiscale entropy profile. Contrary to multiscale entropy, multiscale entropy scoring can be followed as a function of time to monitor disease progression with limited loss of information. Analysis of multiscale complexity of beat-to-beat dynamics at high temporal resolution has potential as a sensitive prognostic tool with translational power that can predict survival outcome in systemic inflammatory conditions such as sepsis and septic shock.
Product Description
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Octoclothepin maleate salt, solid