Merck
  • The Park Grass Experiment and next-generation approaches: local adaptation of sweet vernal grass revisited.
The Park Grass Experiment and next-generation approaches: local adaptation of sweet vernal grass revisited.
Molecular ecology (2014-12-24)
Eric J B von Wettberg, Wendy Vance, Diane L Rowland
ABSTRACT
Long-term ecological experiments provide unique opportunities to observe the effects of natural selection. The Park Grass Experiment at Rothamsted Experiment Station in Hertfordshire, UK, is the longest running ecological experiment that incorporates fertilization treatments and has been ongoing since 1856. In the 1970s, local adaptation was observed in the grass Anthoxanthum odoratum to the elevated soil aluminium levels of the fertilized plots. Gould et al. (2014) have utilized this system to reevaluate the extent of local adaptation, first documented nearly 45 years ago (Snaydon), and to use emerging molecular approaches to identify candidate genes for the adaptation. From their work, they identify several plausible candidate loci for aluminium tolerance. This work shows the power of long-term field-based trials in a scientific age concentrated on rapidly emerging molecular techniques often utilized in short, narrowly focused laboratory or controlled environment experiments. The current study clearly illustrates the benefits gained by combining these molecular approaches within long-term monitoring experiments that can be regularly revisited in a changing world and used to address questions on evolutionary scales.
Product Description
Aluminum, wire reel, 5m, diameter 0.25mm, as drawn, 99.5%
Aluminum, wire reel, 50m, diameter 0.25mm, as drawn, 99.95+%
Aluminum, wire reel, 50m, diameter 0.125mm, as drawn, 99.5%
Aluminum, wire reel, 50m, diameter 1.5mm, as drawn, 99.999%
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 16 pores/cm, aluminum 6101 grade, 98.5%
Aluminum, foam, 150x150mm, thickness 3.2mm, bulk density 0.2g/cm3, porosity 0.93, 16 pores/cm, aluminum 6101 grade, 98.5%
Aluminum, honeycomb, 600x600mm, thickness 10mm, cell size 3.2mm, cell wall 0.025mm, core density 0g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, honeycomb, 600x600mm, thickness 26.5mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin glass fiber 0.5mm, 99.9%
Aluminum, honeycomb, 300x300mm, thickness 20mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, honeycomb, 150x150mm, thickness 10mm, cell size 3.2mm, cell wall 0.025mm, core density 0g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, foam, 150x150mm, thickness 10mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Aluminum, honeycomb, 600x600mm, thickness 13.9mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin aluminum 0.55mm, 99.9%
Aluminum, honeycomb, 600x600mm, thickness 26.5mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin aluminum 0.55mm, 99.9%
Aluminum, honeycomb, 300x300mm, thickness 26.5mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin glass fiber 0.5mm, 99.9%
Aluminum, honeycomb, 300x300mm, thickness 10mm, cell size 3.2mm, cell wall 0.025mm, core density 0g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, foam, 33x140mm, thickness 20mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Aluminum, honeycomb, 600x600mm, thickness 20mm, cell size 4.8mm, cell wall 0.038mm, core density 0.070g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 4 pores/cm, aluminum 6101 grade, 98.5%
Aluminum, honeycomb, 300x300mm, thickness 5mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, wire reel, 0.2m, diameter 3.0mm, as drawn, 99.999%
Aluminum, wire reel, 100m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 1000m, diameter 1.0mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.5mm, as drawn, 99.5%
Aluminum, wire reel, 0.5m, diameter 1.5mm, as drawn, 99.999%