Merck
  • An aberrant leukotriene A4 hydrolase-proline-glycine-proline pathway in the pathogenesis of chronic obstructive pulmonary disease.
An aberrant leukotriene A4 hydrolase-proline-glycine-proline pathway in the pathogenesis of chronic obstructive pulmonary disease.
American journal of respiratory and critical care medicine (2014-05-31)
J Michael Wells, Philip J O'Reilly, Tomasz Szul, Daniel I Sullivan, Guy Handley, Chris Garrett, Carmel M McNicholas, Mojtaba Abdul Roda, Bruce E Miller, Ruth Tal-Singer, Amit Gaggar, Stephen I Rennard, Patricia L Jackson, J Edwin Blalock
ABSTRACT
Chronic neutrophilic inflammation is a hallmark in the pathogenesis of chronic obstructive pulmonary disease (COPD) and persists after cigarette smoking has stopped. Mechanisms involved in this ongoing inflammatory response have not been delineated. We investigated changes to the leukotriene A4 hydrolase (LTA4H)-proline-glycine-proline (PGP) pathway and chronic inflammation in the development of COPD. A/J mice were exposed to air or cigarette smoke for 22 weeks followed by bronchoalveolar lavage and lung and cardiac tissue analysis. Two human cohorts were used to analyze changes to the LTA4H-PGP pathway in never smokers, control smokers, COPD smokers, and COPD former smokers. PGP/AcPGP and LTA4H aminopeptidase activity were detected by mass spectroscopy, LTA4H amounts were detected by ELISA, and acrolein was detected by Western blot. Mice exposed to cigarette smoke developed emphysema with increased PGP, neutrophilic inflammation, and selective inhibition of LTA4H aminopeptidase, which ordinarily degrades PGP. We recapitulated these findings in smokers with and without COPD. PGP and AcPGP are closely associated with cigarette smoke use. Once chronic inflammation is established, changes to LTA4H aminopeptidase remain, even in the absence of ongoing cigarette use. Acrolein modifies LTA4H and inhibits aminopeptidase activity to the same extent as cigarette smoke. These results demonstrate a novel pathway of aberrant regulation of PGP/AcPGP, suggesting this inflammatory pathway may be intimately involved in disease progression in the absence of ongoing cigarette smoke exposure. We highlight a mechanism by which acrolein potentiates neutrophilic inflammation through selective inhibition of LTA4H aminopeptidase activity. Clinical trial registered with www.clinicaltrials.gov (NCT 00292552).
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
SAFC
L-Proline
SAFC
Glycine
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Proline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine 1 M solution
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)