Merck
  • Biotin-responsive basal ganglia disease should be renamed biotin-thiamine-responsive basal ganglia disease: a retrospective review of the clinical, radiological and molecular findings of 18 new cases.
Biotin-responsive basal ganglia disease should be renamed biotin-thiamine-responsive basal ganglia disease: a retrospective review of the clinical, radiological and molecular findings of 18 new cases.
Orphanet journal of rare diseases (2013-06-08)
Majid Alfadhel, Makki Almuntashri, Raafat H Jadah, Fahad A Bashiri, Muhammad Talal Al Rifai, Hisham Al Shalaan, Mohammed Al Balwi, Ahmed Al Rumayan, Wafaa Eyaid, Waleed Al-Twaijri
ABSTRACT
Biotin-responsive basal ganglia disease (BBGD) is an autosomal recessive neurometabolic disorder. It is characterized by sub acute encephalopathy with confusion, seizure, dysarthria and dystonia following a history of febrile illness. If left untreated with biotin, the disease can progress to severe quadriparesis and even death. A retrospective chart review of 18 patients with BBGD from two tertiary institutions describing their clinical, magnetic resonance imaging and molecular findings was conducted. Eighteen children from 13 families seen over a period of nine years (2003-2012) were included. (Age range: 14month to 23 years, M: F: 1:1). The clinical features included sub acute encephalopathy, ataxia (n= 18), seizures (n= 13) dystonia (n=12) ,dysarthria (n= 9), quadriparesis and hyperreflexia (n=9). Magnetic resonance imaging demonstrated abnormal signal intensity with swelling in the basal ganglia during acute crises (n= 13/13) and atrophy of the basal ganglia and necrosis during follow up (n= 13/13). One-third of the present patients showed the recurrence of acute crises while on biotin therapy alone, but after the addition of thiamine, crises did not recur. All of the patients have a homozygous missense mutation in exon 5 of the SLC19A3 gene. The frequency of acute crises, delay in diagnosis and initiation of treatment significantly influenced the outcome. On follow up, four patients died, two had spastic quadriplegia, six had normal outcome and the rest had speech and motor dysfunctions. Clinicians should suspect BBGD in any child presenting with sub acute encephalopathy, abnormal movement and MRI findings as described above. Both biotin and thiamine are essential for disease management. Since biotin alone could not prevent the recurrence of crises in some patients, a more appropriate term to describe the disease would be biotin-thiamine-responsive basal ganglia disease (BTBGD).
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, ≥99.0% (T)
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Biotin, European Pharmacopoeia (EP) Reference Standard
SAFC
BIOTIN
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Biotin, United States Pharmacopeia (USP) Reference Standard