Merck
  • Evaluation and management of acquired methemoglobinemia associated with topical benzocaine use.
Evaluation and management of acquired methemoglobinemia associated with topical benzocaine use.
American journal of cardiovascular drugs : drugs, devices, and other interventions (2013-05-23)
Mohammed Taleb, Zubair Ashraf, Shahul Valavoor, Jodi Tinkel
ABSTRACT
Benzocaine is a widely used topical oropharyngeal anesthetic and has been reported to cause methemoglobinemia. We discuss benzocaine-induced methemoglobinemia and review the causes, presentation, and management of this serious complication. Treatment with methylene blue will result in reversal of methemoglobinemia and clinical recovery in most cases but needs to be used at appropriate doses in carefully selected individuals. Physicians who perform procedures involving the application of benzocaine for topical anesthesia need to rapidly identify and treat methemoglobinemia to avoid significant associated morbidity and mortality.
Product Description
Supelco
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Benzocaine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%