Merck
  • Decontamination of textile wastewater via TiO2/activated carbon composite materials.
Decontamination of textile wastewater via TiO2/activated carbon composite materials.
Advances in colloid and interface science (2010-08-03)
K Y Foo, B H Hameed
ABSTRACT
Water scarcity and pollution rank equal to climate change as the most urgent environmental turmoil for the 21st century. To date, the percolation of textile effluents into the waterways and aquifer systems, remain an intricate conundrum abroad the nations. With the renaissance of activated carbon, there has been a steadily growing interest in the research field. Recently, the adoption of titanium dioxide, a prestigious advanced photo-catalyst which formulates the new growing branch of activated carbon composites for enhancement of adsorption rate and discoloration capacity, has attracted stern consideration and supports worldwide. Confirming the assertion, this paper presents a state of art review of titanium dioxide/activated carbon composites technology, its fundamental background studies, and environmental implications. Moreover, its major challenges together with the future expectation are summarized and discussed. Conclusively, the expanding of activated carbons composites material represents a potentially viable and powerful tool, leading to the plausible improvement of environmental conservation.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Supelco
Activated charcoal, powder
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%
Carbon, foil, not light tested, 150x150mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 1.5mm, graphite, 100%
Carbon, rod, 150mm, diameter 13.0mm, graphite, 100%
Carbon, microleaf, 25x70mm, thinness 0.050μm, specific density 10μg/cm2, temporary glass support, 99.997%
Carbon, tube, graphite, 100mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Carbon, rod, 300mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon, rod, 50mm, diameter 25mm, graphite, 100%
Carbon, microleaf, 25x70mm, thinness 0.25μm, specific density 50μg/cm2, temporary glass support, 99.997%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon, tube, graphite, 100mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 100mm, diameter 2.0mm, graphite, 100%
Carbon, rod, 300mm, diameter 13.0mm, graphite, 100%
Carbon, foil, 50x50mm, thickness 0.35mm, flexible graphite, 99.8%