Merck
Impact of silver nanoparticles and silver ions on innate immune cells.
Journal of biomedical nanotechnology (2014-04-23)
Hajo Haase, Amir Fahmi, Boris Mahltig
ABSTRACT
Silver is commonly used as an antibacterial agent, e.g., in various medical applications, and the availability of silver nanoparticles (AgNP) has fueled this development. Their antibacterial properties are well defined, whereas there are concerns regarding unknown and potentially harmful effects of AgNPs on immune cells and an ongoing immune reaction. Aim of the present study is a comparison of the effects of AgNPs and ionic silver (Ag+) on cells of the innate immune system, in particular on neutrophil granulocytes and macrophages. The AgNPs were synthesized within hydroxylated polyester dendrimer templates via an in situ approach, generating five kinds of AgNPs with mean diameters from 2.0 to 34.7 nm.4 No impact is observed on phagocytosis and oxidative burst, as well as activation of the promoter for the pro-inflammatory cytokine TNF-alpha. In contrast, both AgNPs and Ag+, but not the dendrimer templates, trigger the release of neutrophil extracellular traps and inhibit the formation of nitric monoxide. On the molecular level, AgNPs and Ag+ cause elevated intracellular levels of reactive oxygen species and the second messenger Zn2+. Moreover, protein phosphatases are inhibited by an oxidative mechanism. Taken together, there are several effects of AgNPs on neutrophil granulocytes and macrophages in vitro, but these are not specific for AgNP, instead they are also observed with Ag+, and Ag+ released from AgNPs seems to be the component responsible for most of the particles' immunomodulatory activity.
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%
Silver, foil, 6mm disks, thickness 0.008mm, 99.97%
Silver, foil, 8mm disks, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, light tested, 300x300mm, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.005mm, 99.97%
Silver, foil, not light tested, 150x150mm, thickness 0.01mm, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.02mm, 99.97%
Silver, foil, not light tested, 100x100mm, thickness 0.003mm, 99.9%
Silver, foil, 8mm disks, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, light tested, 50x50mm, thickness 0.0125mm, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.0025mm, 99.97%
Silver, foil, 1m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.01mm, 99.95+%
Silver, foil, 1m coil, thickness 0.01mm, 99.95+%
Silver, foil, 15mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Silver, foil, 15mm disks, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.05mm, annealed, 99.95+%
Silver, foil, 15mm disks, thickness 0.025mm, as rolled, 99.99+%