Skip to Content
Merck
All Photos(2)

Key Documents

GF37871792

Silver

foil, light tested, 100x100mm, thickness 0.02mm, 99.97%

Synonym(s):

Silver, AG000190

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,430.00

$2,430.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$2,430.00

About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

$2,430.00


Check Cart for Availability

Request a Bulk Order

Assay

99.97%

form

foil

manufacturer/tradename

Goodfellow 378-717-92

resistivity

1.59 μΩ-cm, 20°C

size × thickness

100x100 mm × 0.02 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugene A Chu et al.
Otology & neurotology : official publication of the American Otological Society, American Neurotology Society [and] European Academy of Otology and Neurotology, 24(3), 507-518 (2003-06-14)
To present the rich and checkered history of the artificial eardrum, a widely used device in the 19th century, and to illustrate the behavior of otologists in response to the introduction of a promising new technology. Over 40 published books
Fan Yang et al.
Talanta, 84(4), 1099-1106 (2011-05-03)
In this paper, a compact and inexpensive light emitting diode induced fluorescence (LED-IF) detector with simplified optical configuration was developed and assembled in an integrated microfluidic device for microscale electrophoresis. The facile detector mainly consisted of an LED, a focusing
G J Hoffman et al.
Science (New York, N.Y.), 177(4051), 802-804 (1972-09-01)
The water potential of leaves in situ can be measured without temperature control with a miniature, single-junction psychrometer constructed from silver foil and attached to the leaf with a silver-impregnated, conductive coating. The temperature of the psychrometer has been found
D C Sharma et al.
Indian journal of physiology and pharmacology, 41(3), 285-288 (1997-07-01)
About 50 mg of silver leaf (metallic silver) was given daily by mouth to 30 healthy volunteers for 20 days. A statistically significant hypophospholipidemic, hypotriglyceridemic, hypocholesterolemic and hypoglycemic effect was observed. This was accompanied by a less marked fall in
A Rupérez et al.
Talanta, 44(2), 213-220 (1997-02-01)
Surface-enhanced Raman scattering (SERS) spectrometry of the diuretic drug triamterene is discussed. The SERS-active substrate used is a silver foil etched with nitric acid. The influence of solvent and sample doping method on sensivity, intercept and shape of the calibration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service