Merck
  • Novel hypoglycemic injury mechanism: N-methyl-D-aspartate receptor-mediated white matter damage.
Novel hypoglycemic injury mechanism: N-methyl-D-aspartate receptor-mediated white matter damage.
Annals of neurology (2013-11-19)
Xin Yang, Margaret A Hamner, Angus M Brown, Richard D Evans, Zu-Cheng Ye, Shengdi Chen, Bruce R Ransom
ABSTRACT
Hypoglycemia is a common adverse event and can injure central nervous system (CNS) white matter (WM). We determined whether glutamate receptors were involved in hypoglycemic WM injury. Mouse optic nerves (MON), CNS WM tracts, were maintained at 37°C with oxygenated artificial cerebrospinal fluid (ACSF) containing 10mM glucose. Aglycemia was produced by switching to 0 glucose ACSF. Supramaximal compound action potentials (CAPs) were elicited using suction electrodes, and axon function was quantified as the area under the CAP. Amino acid release was measured using high-performance liquid chromatography. Extracellular lactate concentration ([lactate(-)]o) was measured using an enzyme electrode. About 50% of MON axons were injured after 60 minutes of aglycemia (90% after 90 minutes); injury extent was not affected by animal age. Blockade of N-methyl-D-aspartate (NMDA)-type glutamate receptors improved recovery after 90 minutes of aglycemia by 250%. Aglycemic injury was increased by reducing [Mg(2+)]o or increasing [glycine]o , and decreased by lowering pHo , expected results for NMDA receptor-mediated injury. pHo increased during aglycemia due to a drop in [lactate(-)]o. Aglycemic injury was dramatically reduced in the absence of [Ca(2+)]o. Extracellular aspartate, a selective NMDA receptor agonist, increased during aglycemia ([glutamate]o fell). Aglycemia injured WM by a unique excitotoxic mechanism involving NMDA receptors (located primarily on oligodendrocytes). During WM aglycemia, the selective NMDA agonist aspartate is released, probably from astrocytes. Injury is mediated by Ca(2+) influx through aspartate-activated NMDA receptors made permeable by an accompanying alkaline shift in pHo caused by a fall in [lactate(-)]o. These insights have important clinical implications.
Sigma-Aldrich
Glycogen from bovine liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
(±)-AMPA, solid
Sigma-Aldrich
NBQX disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Glycogen from oyster
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Glycine 1 M solution
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, for DNA precipitations
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, ≥85% anhydrous basis (enzymatic)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
NBQX hydrate, powder, ≥98% (HPLC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)