Merck
  • Cellular prion protein (PrPC) of the neuron cell transformed to a PK-resistant protein under oxidative stress, comprising main mitochondrial damage in prion diseases.
Cellular prion protein (PrPC) of the neuron cell transformed to a PK-resistant protein under oxidative stress, comprising main mitochondrial damage in prion diseases.
Journal of molecular neuroscience : MN (2013-05-30)
Fangzhong Yuan, Lifeng Yang, Zhuming Zhang, Wenyu Wu, Xiangmei Zhou, Xiaomin Yin, Deming Zhao
ABSTRACT
Prion diseases characterize a category of fatal neurodegenerative diseases. Although reports have increasingly shown that oxidative stress plays an important role in the progression of prion diseases, little is known about whether oxidative stress is a cause or a consequence of a prion disease. The mechanism of prion disease development also remains unclear. The purpose of this study was to investigate three things: the possible mechanisms of neuron cell damage, the conformation of anti-protease K (PK) PrP(Sc), and the role of oxidative stress in the progression of prion diseases. The study results demonstrated that normal PrP(C) transformed into a PK-resistant protein under oxidative stress in the presence of PrP106-126. Further, the protein misfolding cyclic amplification procedure may have accelerated this process. Mitochondrial damage and dysfunction in prion disease progression were also observed in this study. Our results suggested that neuron cell damage, and particularly mitochondrial damage, was induced by oxidative stress. This damage may be the initial cause of a given prion disease.
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥3.0 unit/mg solid, lyophilized powder
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Proteinase K from Tritirachium album, Reagents designed and manufactured under current ISO 13485 certification.