Merck
  • Development of sustained-release matrix tablets of BKP-01-041 (tilorone derivative) containing Hypromellose.
Development of sustained-release matrix tablets of BKP-01-041 (tilorone derivative) containing Hypromellose.
Die Pharmazie (2013-11-28)
Liangmei Chen, Fei Wang
ABSTRACT
The objective of this research was to develop and evaluate sustained-release matrix tablets of BKP-01-041 (tilorone derivative) based on Hypromellose (hydroxypropyl methylcellulose, HPMC) as the matrix forming polymer. The sustained-release tablets were prepared by the wet granulation method. The influence of HPMC viscosity and ratios on drug release was investigated in vitro. Dissolution of the tablets developed with 26% HPMC K4 M/K100 M (1:2) (w/w) content showed a better drug release profile than the other batches tested in 12 h. Drug release from the optimal formulation was analyzed using release kinetics equations. The release kinetics parameters were determined and the value of the exponent (n) representing the apparent drug release mechanism determined from the Peppas equation was about 0.726. These results suggest that the drug release mechanism was non-Fickian (0.45 < n < 0.89), and drug release was dependent on both drug diffusion and polymer erosion.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Tilorone dihydrochloride, 95%
Sigma-Aldrich
Methyl cellulose, Vetec, reagent grade