Skip to Content
Merck
All Photos(2)

Key Documents

V900506

Sigma-Aldrich

Methyl cellulose

Vetec, reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shi-You Jiang et al.
Nature communications, 9(1), 5138-5138 (2018-12-05)
Statins are inhibitors of HMG-CoA reductase, the rate-limiting enzyme of cholesterol biosynthesis, and have been clinically used to treat cardiovascular disease. However, a paradoxical increase of reductase protein following statin treatment may attenuate the effect and increase the side effects.
Matthew J Caicco et al.
Journal of controlled release : official journal of the Controlled Release Society, 166(3), 197-202 (2013-01-12)
Stimulation of endogenous neural stem/progenitor cells (NSPCs) with therapeutic factors holds potential for the treatment of stroke. Cyclosporin A (CsA) is a particularly promising candidate molecule because it has been shown to act as a survival factor for these cells
Yihua Wang et al.
Biochemistry, 52(9), 1611-1621 (2013-02-07)
Myosin powers contraction in heart and skeletal muscle and is a leading target for mutations implicated in inheritable muscle diseases. During contraction, myosin transduces ATP free energy into the work of muscle shortening against resisting force. Muscle shortening involves relative
Hisham Al-Obaidi et al.
International journal of pharmaceutics, 443(1-2), 95-102 (2013-01-10)
Solid dispersions of varying weight ratios compositions of the nonionic drug, griseofulvin and the hydrophilic, anionic polymer, hydroxylpropylmethyl cellulose acetate succinate, have been prepared by ball milling and the resulting samples characterized using a combination of Fourier transform infra-red spectroscopy
Eyal Zur
International journal of pharmaceutical compounding, 17(1), 10-22 (2013-05-01)
Compounding pharmacists throughout the world are compounding a special type of capsule called "slow-release." This type of capsule is a compounding pharmacy application of the commercial hydrophilic matrix tablets. It is a relatively simple system that allows formulating a robust

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service