Merck
  • Optimization of sustained release matrix tablet of metoprolol succinate using central composite design.
Optimization of sustained release matrix tablet of metoprolol succinate using central composite design.
Pakistan journal of pharmaceutical sciences (2013-09-17)
Li Li, Huijuan Sun, Jing Gao, Tao Jiang, Yuan Gao, Jianjun Zhang
ABSTRACT
The present study was performed to optimize the formulation of metoprolol succinate (MS) sustained release tablets using hydroxypropyl methylcellulose (HPMC) and sodium alginate (SA) as the matrix combination. After investigating the effects of various parameters on drug release, a 2-factor, 5-level central composite design was employed, using the amount of HPMC K4M (A) and SA (318 cP) (B) as the independent variables and the drug percentage released at 1h, 4h, 8h, 20h (Q1, Q4, Q8, Q20) as the responses. Response surfaces were established to obtain the matrix ranges and the main factors affecting four responses. In order to validate the optimization study, six confirmatory runs were performed; indicating high predictability of response surface methodology for MS sustained release tablets. Data fitting to Peppas equation indicated that the mechanism of drug release could be diffusion along with erosion. This matrix combination can be used as a good alternative to the commercially pellet technology, which was complicated, time-consuming and energy-intensive.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(±)-Metoprolol (+)-tartrate salt, ≥98% (titration), powder
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Methyl cellulose, Vetec, reagent grade
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Alginic acid from brown algae, powder
Metoprolol tartrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Metoprolol Tartrate, Pharmaceutical Secondary Standard; Certified Reference Material