Merck
  • Baicalein and hydroxypropyl-γ-cyclodextrin complex in poloxamer thermal sensitive hydrogel for vaginal administration.
Baicalein and hydroxypropyl-γ-cyclodextrin complex in poloxamer thermal sensitive hydrogel for vaginal administration.
International journal of pharmaceutics (2013-07-16)
Qiuna Zhou, Lin Zhong, Xiaohui Wei, Wei Dou, Guixin Chou, Zhengtao Wang
ABSTRACT
The present study aimed to prepare a chemically and physically stable formulation of baicalein (Ba) in an in situ thermally sensitive hydrogel for vaginal administration. An inclusion complex of Ba and hydroxypropyl-γ-cyclodextrin (HP-γ-CD) was first developed to increase the stability and solubility of Ba in an aqueous solution. The formation of the Ba-HP-γ-CD complex was characterized by scanning electron microscopy (SEM), differential scanning calorimetry (DSC), and X-ray powder diffractometry (XRPD). Poloxamers 407, 188 (P407, P188), sodium alginate (SA), hydroxypropylmethylcellulose (HPMC), and benzalkonium bromide were mixed to obtain a hydrogel with an appropriate gelation temperature for vaginal use. Among the formulations examined, P407/Ba-HP-γ-CD/HPMC/P188/benzalkonium bromide (18/0.96/0.5/4.0/0.02%) represented the appropriate gelation temperature and acceptable drug release rate at the administration site. Release of Ba-HP-γ-CD from the poloxamer hydrogel followed Peppas equation, which suggests that it occurred through coupled corrosion-diffusion mechanism, and the corrosion-release curve showed that corrosion was the primary mode of release. In animal study, treatment by using Ba-HP-γ-CD thermosensitive hydrogel could produce a restoration of damaged tissues. Thermosensitive hydrogel formulation of the Ba-HP-γ-CD complex appears to be a promising treatment for cervicitis.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, Vetec, reagent grade
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Alginic acid from brown algae, powder
Sigma-Aldrich
Sodium alginate