Merck
  • Blood methanethiol in alcoholic liver disease with and without hepatic encephalopathy.
Blood methanethiol in alcoholic liver disease with and without hepatic encephalopathy.
Gut (1980-04-01)
C J McClain, L Zieve, W M Doizaki, S Gilberstadt, G R Onstad
ABSTRACT
Blood methanethiol and ammonia concentrations were measured in 16 healty volunteers, 52 consecutive alcoholic cirrhotics without overt hepatic encephalopathy (HE), and 42 consecutive patients with alcoholic liver disease and overt HE. The mean concentration of blood methanethiol was significantly greater than normal in the cirrhotics without overt HE, and the means of both methanethiol and ammonia were significantly greater in the patients with than in those without overt HE. Only one patient with overt HE had both normal ammonia and methanethiol blood concentrations. Twenty of the patients with HE were followed serially. The directions of change in methanethiol and ammonia were consistent with the direction of change in mental status in 85% adn 60% respectively. All of the patients who deteriorated and died had changes in blood methanethiol that correlated with the change in mental status. We conclude that blood methanethiol is a valuable adjunct to the ammonia determination in the evaluation of the patient with possible HE. It is especially helpful in following the course of a patient with hepatic encephalopathy, both as to prognosis and as an indicator of response to therapy.
Product Description
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Methanethiol, purum
SAFC
L-Aspartic acid
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
2,3-Butanediol, 98%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Tyramine, 98%, FG
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
Urea, meets USP testing specifications
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)