Merck
  • Parenteral nutrition-associated cholestasis in neonates: the role of aluminum.
Parenteral nutrition-associated cholestasis in neonates: the role of aluminum.
Nutrition reviews (2003-10-14)
Chris J Arnold, Grant G Miller, Gordon A Zello
ABSTRACT
Parenteral nutrition (PN) is an essential component in the care of premature and ill infants. The incidence of parenteral nutrition-associated cholestasis (PNAC) ranges from 7.4 to 84%. One substance in PN solutions that has been implicated in PNAC is aluminum. Aluminum loading in animals and humans causes hepatic accumulation and damage. The degree of aluminum contamination of PN solutions has decreased over time, but contamination still significantly exceeds levels that are safe for human neonates. Further study into the relationship between aluminum contamination in neonatal PN solutions and the development of PNAC is necessary.
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil