Merck
1H and (13)C NMR spectroscopic analysis of human saliva.
Journal of dental research (2002-07-05)
C J L Silwood, E Lynch, A W D Claxson, M C Grootveld
ABSTRACT
We have explored the ability of high-resolution NMR techniques to (1) index salivary biomolecules and (2) provide valuable data regarding intra- and inter-subject variability in the concentrations of a series of components readily determinable by this technique (organic acids and malodorous amines). Experiments were conducted on 'whole' saliva samples collected from 20 patients, either randomly during their daily activities, or, for investigations involving the quantification of salivary biomolecules, immediately after they woke in the morning throughout a three-day period. These NMR techniques permitted us to detect greater than 60 metabolites, together with agents arising from dietary, oral health care product, and pharmaceutical sources. Highly significant "between-subject" differences in the a.m. waking salivary metabolite concentrations were found for 9 out of 11 components monitored. It is concluded that NMR spectroscopy serves as a powerful technique for the multicomponent analysis of human saliva.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
L-Alanine, ≥99%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG