Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Phytochemical, antioxidant and protective effect of Rhus tripartitum root bark extract against ethanol-induced ulcer in rats.
Phytochemical, antioxidant and protective effect of Rhus tripartitum root bark extract against ethanol-induced ulcer in rats.
General physiology and biophysics (2013-03-28)
Hichem Alimi, Sakhria Mbarki, Zeineb B Barka, Anwer Feriani, Zouhour Bouoni, Najla Hfaeidh, Mohsen Sakly, Olfa Tebourbi, Khémais B Rhouma
ABSTRACT
Rhus tripartitum (sumac) is an Anacardiaceae tree with a wide phytotherapeutic application including the use of its roots in the management of gastric ulcer. In the present study the Rhus tripartitum root barks extract (RTE) was phytochemical studied, in vitro tested for their potential antioxidant activity using 2,2-diphenyl-1-picrylhydrazyl (DPPH) and reducing power assay and in vivo evaluated for its ability to prevent ethanol-induced gastric ulcer in rats. The RTE was rich in phenolics, flavonoids, tannins and polysaccharide contents and exhibited a low but not weak in vitro antioxidant activity when compared with (+)-catechin. Pre-treatment with RTE at oral doses 50, 200 and 400 mg/kg body weight was found to provide a dose-dependent protection against ethanol-induced ulcer by averting the deep ulcer lesions of the gastric epithelium, by reducing gastric juice and acid output, by enhancing gastric mucus production by preserving normal antioxidant enzymes activities, and inhibiting the lipid peroxidation. The antiulcerogenic activity of RTE might be due to a possible synergistic antioxidant and antisecretory effects.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)