Merck
  • Prestorage in ovo injection of biological buffers: an approach to improve hatchability in long-term stored eggs.
Prestorage in ovo injection of biological buffers: an approach to improve hatchability in long-term stored eggs.
Poultry science (2013-03-09)
A Akhlaghi, Y Jafari Ahangari, S R Hashemi, B Navidshad, Z Ansari Pirsaraei, H Deldar, M R Ebrahimi, M Dadpasand, H Atashi, J B Liang
ABSTRACT
A hypothesis was tested that the in ovo injection of biological buffers may reinforce the buffering capacity of albumen, thereby withstanding the increase in albumen pH during storage and improving hatchability and chick quality in long-term stored eggs. Hatching eggs (n = 2,420) were randomly assigned to 11 treatment groups (4 replicates of 55 eggs each) and injected (d 1) with distilled water, 25 or 50 mM HEPES (H25 and H50), Bicine (B25 and B50), Tris (T25 and T50), and Bis-Tris-propane (BTP25 and BTP50) solutions or were not injected (intact: control; or pricked with a needle: N). The eggs were then stored for 14 d during which the egg internal characteristics were evaluated at 1, 2, 3, 4, 5, 8, and 13 d of storage (n = 924 in total) and the remaining eggs (n = 1,496) were incubated. A decrease in albumen pH was found for H25, H50, B50, and BTP25 groups from 2 through 5 d postinjection. Eggs receiving H25, H50, and B50 recorded a higher albumen index (at 13 d of storage) and Haugh unit (between 8 and 13 d of storage) compared with the control. Interestingly, the hatchability of fertile eggs was influenced by the treatment effect (P = 0.0001) where the eggs receiving H25 (88.3%), H50 (88.9%), B50 (88.4%), and BTP25 (87.6%) recorded higher values than that of control (82.1%), associated with a decreased early embryonic mortality rate (P < 0.0001). In ovo injection of Tris buffer, however, profoundly decreased the hatchability (47.2 and 29.0% for T25 and T50, respectively) and percentage of first-grade chicks (67.5 and 63.6% for T25 and T50, respectively) compared with the control (90.1%). In conclusion, prestorage in ovo injection of H25, H50, B50, and BTP25 improved hatchability in long-term stored eggs in which a decreased albumen pH during the d 2 through 5 of storage period might be involved.
Product Description
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
Sigma-Aldrich
BIS-TRIS propane, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
BIS-TRIS propane, ≥99.0% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
BIS-TRIS propane, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES sodium salt
SAFC
HEPES
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES sodium salt, Vetec, reagent grade, 96%
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
BICINE, ≥99% (titration)
Sigma-Aldrich
BICINE, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline