Merck
  • Fluorescence spectroscopy of PTCDA molecules on the KCl(100) surface in the limit of low coverages: site selection and diffusion.
Fluorescence spectroscopy of PTCDA molecules on the KCl(100) surface in the limit of low coverages: site selection and diffusion.
Physical chemistry chemical physics : PCCP (2013-02-26)
Alexander Paulheim, Mathias Müller, Christian Marquardt, Moritz Sokolowski
ABSTRACT
We performed fluorescence (FL) and fluorescence excitation (FLE) spectroscopy on the model molecule perylene-3,4,9,10-tetracarboxyl acid dianhydride (PTCDA) for very low coverages (below 1% of a monolayer) on thin (100) oriented KCl films. Two different states of PTCDA molecules can be distinguished in the spectra: an initial state, which is observed directly after deposition of the molecules onto the cold sample at 20 K, and a final state, which is found after intensive optical excitation or thermal annealing of the sample. The spectrum of the final state is blue-shifted with respect to that of the initial state by 130 ± 15 cm(-1) and exhibits lines with significantly reduced widths. This can be explained by diffusion of molecules from initially populated terrace sites to energetically favoured step edge sites. Polarization dependent spectroscopy reveals the same azimuthal orientation of the molecules on both adsorption sites and leads to a model of the adsorption geometry of PTCDA at the KCl step sites. Our experiment demonstrates how optical spectroscopy can be used to investigate kinetic processes of fluorescent molecules on surfaces.
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%