Merck
  • Effect of drug-polymer interactions on the aqueous solubility of milled solid dispersions.
Effect of drug-polymer interactions on the aqueous solubility of milled solid dispersions.
International journal of pharmaceutics (2013-02-16)
Hisham Al-Obaidi, M Jayne Lawrence, Sonal Shah, Henna Moghul, Noor Al-Saden, Fiza Bari
ABSTRACT
The role of molecular interactions in ball milled solid dispersions in determining the aqueous solubility of the poorly water-soluble drug, griseofulvin (GF) has been examined. Ball milled solid dispersions of GF and hydroxypropylmethylcellulose acetate succinate (HPMCAS) and GF and polyvinylpyrrolidone (PVP) were prepared and characterized by laser diffraction, scanning electron microscopy and X-ray powder diffraction and the aqueous saturation solubility measured and analyzed using one way ANOVA. The results showed that solid dispersions of GF and HPMCAS possessed an aqueous GF saturation solubility of about ten times higher than the GF solubility achieved from PVP-based solid dispersions. Furthermore, although the aqueous solubility of GF did not vary with the milling conditions used to prepare the solid dispersions with PVP, significant changes in solubility were observed upon changing the milling conditions for preparation of the GF/HPMCAS solid dispersions. Surprisingly, the GF/HPMCAS solid dispersion prepared using spray drying exhibited a significantly lower aqueous solubility than those prepared by bead milling despite their smaller particle size and GF being fully in its amorphous form. It is thought that the higher surface energy of the spray-dried solid dispersions negatively affected the aqueous solubility of GF. In conclusion, the results suggest that the molecular interactions occurring between GF and HPMCAS affect the aqueous solubility of GF and that the molecular interactions appear to remain in the liquid state. In contrast no molecular interactions were evident in the GF/PVP solid dispersions.
Product Description
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methyl cellulose, Vetec, reagent grade
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)