Merck
  • NOD-scid IL2R γnull mice engrafted with human peripheral blood mononuclear cells as a model to test therapeutics targeting human signaling pathways.
NOD-scid IL2R γnull mice engrafted with human peripheral blood mononuclear cells as a model to test therapeutics targeting human signaling pathways.
Journal of translational medicine (2013-01-09)
Maryam Zadeh-Khorasani, Thomas Nolte, Thomas D Mueller, Markos Pechlivanis, Franziska Rueff, Andreas Wollenberg, Gert Fricker, Eckhard Wolf, Matthias Siebeck, Roswitha Gropp
ABSTRACT
Animal models of human inflammatory diseases have limited predictive quality for human clinical trials for various reasons including species specific activation mechanisms and the immunological background of the animals which markedly differs from the genetically heterogeneous and often aged patient population. Development of an animal model allowing for testing therapeutics targeting pathways involved in the development of Atopic Dermatitis (AD) with better translatability to the patient. NOD-scid IL2R γnull mice engrafted with human peripheral blood mononuclear cells (hPBMC) derived from patients suffering from AD and healthy volunteers were treated with IL-4 and the antagonistic IL-4 variant R121/Y124D (Pitrakinra). Levels of human (h)IgE, amount of B-, T- and plasma- cells and ratio of CD4 : CD8 positive cells served as read out for induction and inhibition of cell proliferation and hIgE secretion. Results were compared to in vitro analysis. hIgE secretion was induced by IL-4 and inhibited by the IL-4 antagonist Pitrakinra in vivo when formulated with methylcellulose. B-cells proliferated in response to IL-4 in vivo; the effect was abrogated by Pitrakinra. IL-4 shifted CD4 : CD8 ratios in vitro and in vivo when hPBMC derived from healthy volunteers were used. Pitrakinra reversed the effect. Human PBMC derived from patients with AD remained inert and engrafted mice reflected the individual responses observed in vitro. NOD-scid IL2R γnull mice engrafted with human PBMC reflect the immunological history of the donors and provide a complementary tool to in vitro studies. Thus, studies in this model might provide data with better translatability from bench to bedside.
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, Vetec, reagent grade