Merck
  • Analysis on structural, SHG efficiency, optical and mechanical properties of KDP single crystals influenced by Glycine doping.
Analysis on structural, SHG efficiency, optical and mechanical properties of KDP single crystals influenced by Glycine doping.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-25)
Mohd Shkir, V Ganesh, N Vijayan, B Riscob, Anoop Kumar, Devendra Kumar Rana, Mohd Shoeb Khan, Mohd Hasmuddin, M A Wahab, R Ramesh Babu, G Bhagavannarayana
ABSTRACT
Good quality single crystals of potassium dihydrogen orthophosphate (KDP) were grown with different doping concentration of Glycine by conventional solution technique in aqueous solution. X-ray diffraction study has been carried out in order to see the effect of dopant on the structural parameters of KDP. There is no additional phase was observed which was further confirmed by Raman spectroscopic analysis. The second harmonic generation efficiency was measured by using Kurtz powder technique. The relative second harmonic generation (SHG) efficiency of the grown crystals was found to be increased with doping concentration up to 2.5 mol%. Optical transmission study also revealed the same behaviour with enhancement up to 2.5 mol% concentration and later decreased but still higher than pure KDP. The mechanical strength was found to increase with increasing the doping concentration.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Potassium phosphate tribasic, Vetec, reagent grade, 97%
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, Vetec, reagent grade, 98%
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis