Merck
  • Synthesis of duloxetine intermediate (S)-3-Chloro-1-(2-thienyl)-1-propanol with liquid-core immobilized Candida pseudotropicalis 104.
Synthesis of duloxetine intermediate (S)-3-Chloro-1-(2-thienyl)-1-propanol with liquid-core immobilized Candida pseudotropicalis 104.
Applied biochemistry and biotechnology (2012-10-17)
Ou Zhimin, Sun Xingyuan, Shi Hanbing, Bi Hongxia
ABSTRACT
(S) -3-Chloro-1-(2-thienyl)-1-propanol was synthesized by the asymmetric reduction of 3-chloro-1-(2-thienyl)propanone with liquid-core immobilized Candida pseudotropicalis 104. The optimum time was 28 h for the re-cultivation of immobilized cells. The optimum film solvent for the liquid-core capsule was 0.3 % chitosan (M (w) 1.0 × 10(5)). Conversion decreased with the increase of the liquid-core capsule diameter and with the addition of more substrates at the same reduction time. The immobilized cells show good reduction ability in a potassium phosphate buffer (pH 6.6~7.2). The material outside the spread speed of immobilized cells was not restricted when the shaking speed was higher than 160 r/min. Liquid-core immobilized cells can be reused 11 times. Compared with the batch reduction, the continuous reduction of 3-chloro-1-(2-thienyl)propanone in the membrane reactor with liquid-core immobilized cells as catalyst can relieve the inhibition from a high-concentration substrate. Conversion and enantiometric excess of (S)-3-chloro-1-(2-thienyl)-1-propanol reached 100 % and >99 % in a continuous reduction of 12 g/L 3-chloro-1-(2-thienyl)propanone for 10 days.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Potassium phosphate tribasic, Vetec, reagent grade, 97%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Potassium phosphate monobasic, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, Vetec, reagent grade, 98%