Merck
  • Small-angle neutron scattering study of temperature vs. salt dependence of clouding in charged micellar system.
Small-angle neutron scattering study of temperature vs. salt dependence of clouding in charged micellar system.
The European physical journal. E, Soft matter (2012-07-06)
A J Chinchalikar, V K Aswal, J Kohlbrecher, A G Wagh
ABSTRACT
Clouding is studied by small-angle neutron scattering (SANS) on a charged micellar system of sodium dodecyl sulphate (SDS) and tetrabutylammonium bromide (TBAB) with varying temperature and salt NaCl. We show that the clouding occurs as a result of increase in the attractive potential between the micelles mediated by the dehydrated TBA(+) counterions on increasing temperature and in the presence of salt. Both micelles and clusters coexist at cloud point temperature (CP) and beyond CP. The addition of salt can be used to obtain CP at room temperature (30° C). The relative effect of different salts on clouding has been found in the order CaCl(2) > MgSO(4) > Na(2)SO(4) > NaF > NaCl > KCl > CsCl > NaBr > NaNO(3). This order is explained on the basis of two important roles played by salt ions: i) counterion condensation that increases the size of the micelles and ii) dehydration of TBA(+) counterions by salt ions for bridging the micelles.
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetrabutylammonium azide
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%